Amplite® Colorimetric Hypochlorite (Hypochlorous Acid) Assay Kit , AAT Bioquest
76482-286
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
:
- Description:Amplite™ Colorimetric hypochlorite (hypochlorous acid) assay kit
- Cat. No.:76482-286
- No. of tests:200
Hypochlorite anion (ClO-) and its protonated form, hypochlorous acid (HClO) are critical reactive oxygen species (ROS) in biological systems.
- In-house QC testing
- Sensitive
- Suggested protocol available
: For research use only.