- Description:Amplite™ Red
- Size:1000 Assays
- Cat. No.:76481-884
Amplite® Red is a sensitive fluorogenic peroxidase substrate that generates a highly red fluorescent product that has maximum absorption of 571 nm and maximum emission of 585 nm.
- In-house QC testing
- Sensitive
- Suggested protocol available