ContactUsLinkComponent
Amplite® IR, AAT Bioquest
Amplite® IR, AAT Bioquest
Catalog # 76481-880
Supplier:  AAT Bioquest
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
undefined
Amplite® IR, AAT Bioquest
Catalog # 76481-880
Supplier:  AAT Bioquest
Supplier Number:  11009
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Amplite™ IR
  • Size:
    1 mg
  • Cat. No.:
    76481-880
Specifications
About this item
Our Amplite® IR is a fluorogenic peroxidase substrate that generates near infrared fluorescence upon reaction with peroxidase and H2O2.
  • In-house QC testing
  • Sensitive
  • Suggested protocol available
Caution: For research use only.