Amplite® Colorimetric Maleimide Quantitation Kit , AAT Bioquest
76481-802
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
:
- Description:Amplite™ colorimetric maleimide quantitation kit
- Cat. No.:76481-802
- No. of tests:100
Maleimides can be directly assayed spectrophotometrically at 302 nm.
- In-house QC testing
- Sensitive
- Suggested protocol available
: For research use only.