ContactUsLinkComponent
Amplite® Blue Peroxidase Substrate, AAT Bioquest
Amplite® Blue Peroxidase Substrate, AAT Bioquest
Catalog # 76481-878
Supplier:  AAT Bioquest
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
undefined
Amplite® Blue Peroxidase Substrate, AAT Bioquest
Catalog # 76481-878
Supplier:  AAT Bioquest
Supplier Number:  11005
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Amplite™ blue
  • Size:
    25 mg
  • Cat. No.:
    76481-878
Specifications
About this item
A variety of substrates are available for detecting peroxidase activity in ELISA assays. Colorimetric HRP substrates (e.g., TMB, OPD and ABTS) have been widely used for years. Each of these substrates varies greatly with respect to its performance characteristics such as detection sensitivity, working range and attainable signal-to-noise ratios. Amplite® Blue peroxidase substrate is a soluble fluorogenic substrate for detection of peroxidase activity. Amplite® Blue allows rapid HRP detection assays to be performed with greater sensitivity than the colorimetric HRP substrates (such as TMB and OPD). The fluorescent product of Amplite® Blue does not photobleach. Amplite® Blue exhibits a flat baseline in assays, which facilitates low-level detection sensitivity and allows for high signal-to-noise ratios.
  • In-house QC testing
  • Sensitive
  • Suggested protocol available
Caution: For research use only.