ContactUsLinkComponent
Amplite® Fluorimetric Butyrylcholinesterase Activity Assay Kit , AAT Bioquest
Amplite® Fluorimetric Butyrylcholinesterase Activity Assay Kit , AAT Bioquest
Catalog # 76485-154
Supplier:  AAT Bioquest
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
undefined
Amplite® Fluorimetric Butyrylcholinesterase Activity Assay Kit , AAT Bioquest
Catalog # 76485-154
Supplier:  AAT Bioquest
Supplier Number:  11407
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Amplite™ fluorimetric butyrylcholinesterase activity assay kit
  • Cat. No.:
    76485-154
  • No. of tests:
    100
Specifications
About this item
Butyrylcholinesterase (EC 3.1.1.8; BChE), also known as pseudocholinesterase or plasma cholinesterase, is mainly synthesized liver and present in blood.
  • In-house QC testing
  • Sensitive
  • Suggested protocol available
Caution: For research use only.