- Description:Amplite™ colorimetric tyrosinase assay kit
- Cat. No.:76485-006
- No. of tests:200
Tyrosinase is of great interest to drug discovery, life science research, food industry and cosmetics industry since it plays an important role in the biosynthetic pathway of melanin.
- In-house QC testing
- Sensitive
- Suggested protocol available