- Description:Amplite™ colorimetric α-ketoglutarate quantitation kit
- Cat. No.:76484-218
- No. of tests:200
Alpha-ketoglutarate (α-Ketoglutarate) is a key molecule in the Krebs cycle determining the overall rate of the citric acid cycle of the organism.
- In-house QC testing
- Sensitive
- Suggested protocol available
This colorimetric α-Ketoglutarate Quantitation Kit offers a sensitive colorimetric assay for quantifying α-ketoglutarate in biological samples.