hBSA *Optimized for Maximum Immunization Response* (10 mg), AAT Bioquest
76481-810
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
- Serum format:Lyophilized
- Sterility:Non sterile
- Description:BSA cationized with hexamethylenediamine
- Size:10 mg
- Storage Temperature:<–20 °C
- Application:Use as a carrier protein for immunization
- Purification:97%, via fractionation
- Cat. No.:76481-810
- Packaging:Bottle
hBSA is the bovine serum albumin modified with hexamethylenediamine to make the proteins bear more amine groups (thus more positive charges at neutral pH).
- Lyophilized powder
- Excellent quality control
- Stronger immune response
- 60000 molecular weight
: For research use only