XFD594 Goat Anti-Rabbit IgG (H+L) *Cross Adsorbed, XFD594 Same Structure to Alexa Fluor™ 594*
Catalog # 76484-560
Supplier: AAT BIOQUEST INC
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
- Antibody Type:Secondary
- Antigen Name:Immunoglobulin G
- Antigen Symbol:IgG
- Clonality:Polyclonal
- Clone:IgG
- Conjugation:Alexa Fluor® 594
- Host:Goat
- Isotype:IgG (H, L)
- Reactivity:Rabbit
- Size:1 mg
- Cross Adsorption:Yes
- Cat. No.:76484-560
- Packaging:Vial
Specifications
About this item
XFD594 is manufactured by AAT Bioquest, and it has the same chemical structure of Alexa Fluor® 594 (Alexa Fluor® is the trademark of ThermoFisher).
- Affinity-purified
- Bright signal
- Cross-adsorbed