ContactUsLinkComponent
HardyDisks™ AST Ofloxacin, 5 µg (OFX-5), Floxin, Hardy Diagnostics
HardyDisks™ AST Ofloxacin, 5 µg (OFX-5), Floxin, Hardy Diagnostics
Catalog # 76523-364
Supplier:  Hardy Diagnostics
CAS Number:  
undefined
HardyDisks™ AST Ofloxacin, 5 µg (OFX-5), Floxin, Hardy Diagnostics
Catalog # 76523-364
Supplier:  Hardy Diagnostics
Supplier Number:  Z8841
CAS Number:  
Specifications
  • Description:
    Antibiotic tests 
  • Cat. No.:
    76523-364
Specifications
About this item
HardyDisk™ AST's are impregnated paper disks used for Antimicrobial Susceptibility Testing (AST), also known as disk diffusion or Kirby-Bauer testing.
Each cartridge contains 50 disks that are spring loaded for smooth and reliable dispensing.