Order Entry
Puerto Rico
ContactUsLinkComponent
 
Saline Lactose Broth, Hardy Diagnostics
  76523-378
 :  
undefined
Saline Lactose Broth, Hardy Diagnostics
  76523-378
 :  U478
 :  

 

  • Pack type:
    PC bottle, 1 L
  • Description:
    Saline lactose broth
  • Size:
    10/pack
  • Minimum Order Quantity:
    5 packs
  • Cat. No.:
    76523-378
  • Fill volume:
    1000 ml

 

 

Recommended for bacterial filter validation studies.

  • Heavy duty bottles are tough and sturdy to reduce breakage
  • Optically clear bottles are made of clarified polycarbonate for clear viewing