Order Entry
Puerto Rico
ContactUsLinkComponent
HHBS [Hanks' Buffer with 20 mM HEPES] (100 ml), AAT Bioquest
HHBS [Hanks' Buffer with 20 mM HEPES] (100 ml), AAT Bioquest
Catalog # 76482-972
Supplier:  AAT Bioquest
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
undefined
HHBS [Hanks' Buffer with 20 mM HEPES] (100 ml), AAT Bioquest
Catalog # 76482-972
Supplier:  AAT Bioquest
Supplier Number:  20011
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    Hank's Balanced Salt Solution with 20 mm Hepes buffer (HHBS)
  • Size:
    100 ml
  • Cat. No.:
    76482-972

Specifications

About this item

Hank's Balanced Salt Solution with 20 mM Hepes buffer (HHBS) is used for a variety of cell culture applications, such as washing cells, transporting cells or tissue samples, diluting cells for counting, and preparing reagents.

  • Excellent buffering capacity
  • Highly cited
  • Protocol available