Order Entry
Puerto Rico
ContactUsLinkComponent
BioClean™ Non Sterile Tie-on Face Mask, Ansell
BioClean™ Non Sterile Tie-on Face Mask, Ansell
Catalog # 76521-430
Supplier:  Ansell Healthcare
undefined
BioClean™ Non Sterile Tie-on Face Mask, Ansell
Catalog # 76521-430
Supplier:  Ansell Healthcare
Supplier Number:  MTA210-0

Specifications

  • Width:
    95 mm
  • Color:
    White
  • Description:
    Tie-on face mask
  • Length:
    210 mm
  • Cat. No.:
    76521-430

Specifications

About this item

Non sterile tie-on face mask.

  • High bacterial and particle efficiency filtration
  • Fully enclosed malleable nose-band
  • Ultrasonically sealed edges
  • Inner and outer facing layer: non-woven polypropylene/polyethylene (hygroscopic)

Providing high particle and bacterial filtration efficiency, the BioClean™ MTA Cleanroom tie-on face mask is manufactured from cleanroom compatible materials to reduce contamination into the controlled environment. The mask features tie-tapes for a secure fastening.

Certifications: ASTM F2101, ISO 2859, ISO 9001:2015.