Formula 409® Cleaner Degreaser Disinfectant, Refill, Essendant
76500-226
- Capacity:128 oz.
- Environmentally Preferable:
- Cat. No.:76500-226
- Packaging:Bottle, refill
Formula 409® Cleaner Degreaser Disinfectant dissolves grease, grime, and food stains while killing 99.9% of germs.
- EPA-registered disinfectant and sanitizer
- Labor-saving formula is ready to go straight from the bottle.
- Works on a wide range of surfaces
This EPA-registered disinfectant and sanitizer is formulated to kill Salmonella, Staph, E. Coli, and athlete's foot fungus on non-food-contact surfaces. From Clorox Commercial Solutions®, this degreaser and disinfectant powers through tough messes on a wide range of surfaces. It is perfect for use in commercial kitchens, schools, and office break rooms. Suitable for use on ceramic, fiberglass, glass, laminates, metal, and plastic.