ContactUsLinkComponent
Clorox® Bleach Germicidal Wipes, Unscented, Essendant
Clorox® Bleach Germicidal Wipes, Unscented, Essendant
Catalog # 76500-136
undefined
Clorox® Bleach Germicidal Wipes, Unscented, Essendant
Catalog # 76500-136
Supplier Number:  CLO30577
Specifications
  • Color:
    White
  • Description:
    Bleach germicidal wipes, unscented
  • Cat. No.:
    76500-136
  • Packaging:
    150/Canister
  • W×L:
    6×5"
Specifications
About this item
Healthcare professionals trust Clorox Healthcare Bleach Germicidal Wipes to help overcome infection control challenges in a constantly changing healthcare environment. Clorox Healthcare Bleach Germicidal clinical wipes are a ready to use, one-step cleaner containing a 1:10 bleach dilution to meet CDC and OSHA guidelines for disinfecting surfaces in healthcare settings.
  • Ready to use cleaner disinfectant; EPA registered to kill C. Difficile and 58 other microorganisms
  • Clean, disinfect, and deodorize with proven efficacy against outbreak-causing viruses like influenza A, influenza B, and norovirus
  • Powerful disinfectant wipes are great for terminal cleaning
  • Three minute contact time to kill C.Diff spores
  • Meets CDC and OSHA recommendations
  • Intended for use in healthcare settings
Designed for daily disinfection, high-touch surfaces, and medical equipment. These wipes are EPA registered to kill a total of 58 microorganisms, including HAI causing bacteria like MRSA and VRE in 30 seconds, outbreak causing viruses such as Influenza A and B viruses, Measles virus and Norovirus in 1 minute, and C. difficile spores, TB, and fungi in 3 minutes. These wipes kill 99.9999% of C. difficile spores, according to the current EPA recommended testing standard (June 2014).
Wipes are engineered to minimize residue and leave less visible residue on surfaces. They are formulated with an anti-corrosion agent and are compatible with common healthcare surfaces including stainless steel, glass, plastic, porcelain, glazed ceramic, laminated surfaces, and vinyl. Intended for use in healthcare settings.