Order Entry
United States
ContactUsLinkComponent
Mr. Clean® Magic Eraser® Roller Mop, Essendant
Mr. Clean® Magic Eraser® Roller Mop, Essendant
Catalog # 76500-098
Supplier:  Avantor
undefined
Mr. Clean® Magic Eraser® Roller Mop, Essendant
Catalog # 76500-098
Supplier:  Avantor

Specifications

  • Description:
    Mr. Clean® Magic Eraser® Roller mop
  • Environmentally Preferable:
  • Cat. No.:
    76500-098

Specifications

About this item

Roller mop uses deep penetrating action to get out grime and dirt trapped in grooves, so floors really shine.

  • Gets out tough, trapped dirt and grime
  • Highly absorbent, reusable mop
  • Mop head size: 10.5×3"
  • Handle length: 45"
  • Blue/white foam head

The highly absorbent, reusable mop rinses clean time after time. Durable metal handle and a wringing lever to keep hands dry. Safe for use on vinyl, tile, marble, and stone.