About this item
Intact Genomics propriety Ig-Fusion™ cloning technology is a simple, rapid and highly efficient cloning kit which allows to directly clone any PCR product(s) to any linearized expression vector at any site.
The PCR fragments can be generated by Intact Genomics high fidelity Pfu DNA polymerase or other high fidelity DNA polymerases, with primers having 15 to 18 bases of homology at their linear ends to where the product need to fuse. The linearized vector can be generated by PCR or restriction enzymes. The kit is so robust that multiple DNA fragments can be assembled simultaneously and cloned into one construct in a single reaction step within short times (usually 10-30 min) with more than 95% cloning efficiency.
The shipping and dry ice packaging cost is $45 for your entire order in the 48 contiguous United States.