About this item
Intact Genomics EHA105 electrocompetent Agrobacterium cells are useful for transgenic operations of rice, tobacco and other plants.
- High transformation efficiencies
- Especially useful for cDNA gDNA library construction
Intact Genomics EHA105 electrocompetent Agrobacterium cells are optimized for the highest transformation efficiencies which is ideal for applications requiring high transformation efficiencies, such as with cDNA or gDNA library construction. The EHA105 strain is useful for transgenic operations of rice, tobacco and other plants. EHA105 contains a rifampicin resistance gene (rif). EHA105 strain also contains an amber basic Ti plasmid pEHA105 (pTiBo542DT-DNA) without self-transport function, which contains the vir gene.
The shipping and dry ice packaging cost is $45 for your entire order in the 48 contiguous United States.