ContactUsLinkComponent
Simple Green d Pro 3 Plus Antibacterial Concentrate, Essendant
Simple Green d Pro 3 Plus Antibacterial Concentrate, Essendant
Catalog # 76500-256
undefined
Simple Green d Pro 3 Plus Antibacterial Concentrate, Essendant
Catalog # 76500-256
Supplier Number:  SMP01001
Specifications
  • Capacity:
    1 gal.
  • Cat. No.:
    76500-256
  • Packaging:
    Bottle
Specifications
About this item
Simple Green d Pro 3 Plus is a hospital-grade antibacterial cleaner that features an extensive kill list of over 75 different bacteria, viruses, and fungi, including antibiotic-resistant superbugs.
  • Kills 99.9% of germs
  • Cleans, disinfects, and eliminates odors
  • Leaves surfaces smelling clean and fresh
  • Concentrated
  • Meets OSHA Bloodborne Pathogen Standard for HIV, HBV and HCV
Simple Green d Pro 3 Plus provides protection against a wide variety of highly contagious and potentially deadly pathogens. The broad-spectrum, bloodborne pathogen compliant formula is proven effective against HIV, MRSA, staph, E. coli, cold, flu, athlete's foot fungus, and more. The non-acidic formula won't harm sealed stone, sealed grout, or glazed tile, making it ideal for all washable, hard, non-porous surfaces. High 1:64 dilution ratios make this antibacterial cleaning concentrate a cost-effective solution for use in virtually any professional facility. Suitable for use in homes, hotels, kitchens, bathrooms, campgrounds, day care centers, classrooms, universities, athletic facilities, food handling and processing areas, office buildings, factories, warehouses, EMS and fire facilities, transportation terminals, medical and dental offices and clinics, labs, veterinary clinics, animal housing facilities, and other animal care facilities. Chemical compound: Quaternary Ammonium Chlorides.