ContactUsLinkComponent
CrossLab Nebulizers for MP-AES, Agilent Technologies
CrossLab Nebulizers for MP-AES, Agilent Technologies
Catalog # AGG8000-70004
undefined
CrossLab Nebulizers for MP-AES, Agilent Technologies
Catalog # AGG8000-70004
Supplier Number:  G8000-70004
Specifications
  • Description:
    Concentric nebulizer for Agilent MP-AES, 40 µm
  • Cat. No.:
    AGG8000-70004
Specifications
About this item
The Agilent OneNeb Series 2 nebulizer offers superior performance with MP-AES instruments.
  • Direct replacement for standard glass concentric nebulizers and other inert nebulizers - that have a 6 mm ext.Ø tip
  • Convenient and flexible, is suitable for use with Agilent ICP-OES and MP-AES systems
  • Compatible with most conventional glass cyclonic and inert spray chambers used with glass concentric nebulizers
  • Improved sensitivity due to the fine aerosol generated by flow blurring nebulization
  • High efficiency at both normal and micro sample uptake rates
  • Greater tolerance to samples with high total dissolved solids (TDS) content;Inert construction ensures the OneNeb can handle most sample matrices
Agilent 4100/4200 Series MP-AES instruments are compatible with a range of Agilent accessories and supplies designed to extend the instrument's capabilities.
The Agilent OneNeb Series 2 nebulizer offers superior performance with MP-AES instruments. Based on flow blurring nebulization technology the OneNeb series 2 nebulizer provides improved sensitivity and greater tolerance to dissolved solids. What's more, the inert construction of the OneNeb nebulizer makes it a universal design, which can handle most sample matrices, including hydrofluoric (HF) acid digests.