CrossLab Inert Spray Chambers for ICP-OES, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Certificates
About this item
ICP-OES inert spray chamber with removable baffle for single and double pass operation.
- Improved Longevity: Durable sandblasted internal finish provides good wettability WITHOUT the requirement for periodic rejuvenation
- Improved Precision: consistent performance as spray chamber ages
- Improved performance with difficult samples: Design optimized to provide fine aerosol for better performance with high TDS samples
- Greater Flexibility: Easily switch between double pass operation (for challenging samples) and single pass operation (for greater sensitivity)
- Just remove or replace the baffle
- Ease of maintenance: Easily assembled and dissassembled without tools
- A Leur slip drain connector allows easy removal/replacement of the drain fitting
- Ideal for Agilent 5100, 5110, 5800, and 5900 ICP-OES and compatible with earlier Agilent 700/Vista and Liberty series instruments with an adapter
The Agilent ICP-OES inert spray chamber is made of polyethylene terephthalate (PET) and has a durable sandblasted internal finish to ensure good drainage. This permenant sandblasted finish allows optimum drainage and continous accurate performance. The spray chamber has a removable internal baffle that allows it to be used as a double or single pass design, essentially two spray chambers in one. Compatible with all Agilent 5100/5110 ICP-OES and 4200/4210 MP-AES instruments. Retrofittable to the earlier Agilent 700/Vista and Liberty Series ICP-OES instruments (with an appropriate adapter).