Order Entry
Puerto Rico
ContactUsLinkComponent
µ-Slide 18-Well Flat Slide, Ibidi
µ-Slide 18-Well Flat Slide, Ibidi
Catalog # MSPP-81821
Supplier:  IBIDI USA MS
µ-Slide 18-Well Flat Slide, Ibidi
Catalog # MSPP-81821
Supplier:  IBIDI USA MS
Supplier Number:  81821

Specifications

  • Volume:
    30 µl (per well)
  • Description:
    18-Well Flat µ-Slide
  • Dimensions:
    25.5×75.5 mm²
  • Cat. No.:
    MSPP-81821
  • Growth area:
    0.2 cm²

Specifications

About this item

A µ-Slide with 18 wells for use in matrix tests and quick immunofluorescence stainings.

  • Low volume screening slide for medium throughput
  • Cost-effective experiments using small numbers of cells and low volumes of reagents
  • Compatible with multi-channel pipettes
  • Coating area: 0.25 cm²

A convenient multi-well plate for quick and easy tests to probe experimental parameters like antibody dilution, seeding density, or most effective drug concentrations.