µ-Slide Angiogenesis Slide, Ibidi
A µ-Slide used to investigate angiogenesis in tube formation assays. Also ideal for 3D cell culture and immunofluorescence staining.
- Complete solution for tube formation experiments
- Requires only a few steps from sample preparation to image analysis
- Brilliant visualization without gel meniscus formation, and with all cells in one focal plane
- Coating area: 0.23 cm² (using 10 µl)
- Well depth: 0.8 mm (inner well)
Cells can be grown on or in gel matrices, e.g. Matrigel™ or directly on the ibidi Polymer Coverslip. Cost-effective experiments require only 10 µl of gel per well. Can be used with a broad range of gels (Matrigel®, collagen, and agarose).