Orders LinkContactUsLinkComponent
µ-Slide Microscopy Rack, Ibidi
µ-Slide Microscopy Rack, Ibidi
µ-Slide microscopy rack, Aluminum, anodized, 127.5×85.5×12 mm
Catalog # MSPP-80030
Marketsource
Supplier:   IBIDI USA MS
Supplier #:  80030
µ-Slide Microscopy Rack, Ibidi
Catalog # MSPP-80030
Supplier:  IBIDI USA MS
Marketsource
µ-Slide Microscopy Rack, Ibidi
µ-Slide microscopy rack, Aluminum, anodized, 127.5×85.5×12 mm
Price
$669.23
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
A microscopy rack for perfect µ-Slide stabilization.
  • Multiwell format is compatible with all inverted microscopes
  • Lid can be fixed with magnetic positioning for additional stabilization
  • Magnetic lids not included, must be ordered separately
The µ-Slide microscopy rack is an aluminum rack made to hold 4 µ-Slides for analysis of multiple samples. The multiwell format is compatible with all inverted microscopes. Rack may be sterilized via autoclave or alcohol. Magnetic lids are sold seperately.
Specifications
  • Description:
    µ-Slide microscopy rack
  • Dimensions:
    127.5×85.5×12 mm
  • Material:
    Aluminum, anodized