Order Entry
Puerto Rico
ContactUsLinkComponent
Coverslips for Chamber Slides, Removable, Ibidi
Coverslips for Chamber Slides, Removable, Ibidi
Catalog # MSPP-10811
Supplier:  IBIDI USA MS
Coverslips for Chamber Slides, Removable, Ibidi
Catalog # MSPP-10811
Supplier:  IBIDI USA MS
Supplier Number:  10811

Specifications

  • Description:
    Glass Coverslips for 3-, 8-, and 12-chamber removable slides
  • L×W:
    60×24 mm
  • Material:
    No. 1.5H, 170 µm (±5 µm) D 263 M Schott glass
  • Cat. No.:
    MSPP-10811

Specifications

About this item

Glass coverslips for 'ibidi Chamber, removable' slides.

  • Ideal for immunofluorescence assays
  • Compatible with the 3-, 8-, and 12-chamber formats
  • Compatible with all mounting media
  • No autofluorescence
  • Low birefringence
Certifications: In compliance with DIN ISO 4796-1 standards.