Epoxy coated tweezers have an impact-resistant surface with excellent flow and good elasticity.
- Good resistance to many diluted acids and alkalis
- Can operate continuously at temperatures up to 120 °C
Compromised of polyester and epoxy resins, plus the electrically conductive additives.