4-Way Collector (The Werner), SCAT
Catalog # 76503-004
Supplier: SCAT AMERICAS, INC.
Specifications
- Description:4-way collector (The Werner) for HPLC rinsing bottle GLS 80, PE-HD electrostatic conductive, 4× GL 45 (m)
- Cat. No.:76503-004
Specifications
About this item
This purging manifold allows you to install up to four SafetyCaps on your purging bottle.
- Safe and easy handling - capillaries are reliably fixed
- Solvent filters have enough space due to the wide GLS 80 opening
- Simply close unused connections with standard bottle closures
The System for Solvent Safety stays permanently closed, the 'safety chain' remains intact during the purging process.