HPLC Supply and Waste Set, V2.0, S.C.A.T.
Catalog # 76503-786
Supplier: SCAT AMERICAS, INC.
Specifications
- Description:HPLC safety set 2.0
- Cat. No.:76503-786
Specifications
About this item
The HPLC Supply and Waste Disposal Set V2.0 is the ultimate and universal set of all required components for solvent supply and waste of all HPLC/UHPLC systems.
- Safety for solvent supply and waste
- Fits every HPLC/UHPLC system
- Including solvent bottles and waste container
- Blocks solvent vapors
- Protects your system from contamination
- Keeps solvent mixtures stable
The safety set reliably connects hoses and blocks hazardous vapors. The set includes all components required for the supply (SafetyCaps, air valves, and laboratory bottles) as well as the utensils required for the disposal (SafetyWasteCap, exhaust filter, and waste canister). In addition, all other components required for individual configuration (fittings, tube connections, blind plugs) are also already included.
Certifications: Fire protection according to UL-94.
Ordering information: All set components can be reordered separately.
Delivery: The set includes:
4× SafetyCap III V2.0 GL 45
4× 1 liter clear glass bottles GL 45
1× 5 liter PE-HD waste container GL 45
1× SafetyWasteCap V2.0 GL 45 (4× connection ∅ 2,3 - 3,2 mm; 1× connection ∅ 6.4 - 9 mm)
10× blind plug for capillary connection
1× angled tube connector 9,5 - 10 mm
2× straight tube connector 6 - 8 mm
1× exhaust filter size M
5× purple fitting Ø 2,3 mm
5× blue fitting Ø 3,2 mm
5× yellow fitting Ø 3,2 mm
5× green fitting Ø 1,6 mm
4× SafetyCap III V2.0 GL 45
4× 1 liter clear glass bottles GL 45
1× 5 liter PE-HD waste container GL 45
1× SafetyWasteCap V2.0 GL 45 (4× connection ∅ 2,3 - 3,2 mm; 1× connection ∅ 6.4 - 9 mm)
10× blind plug for capillary connection
1× angled tube connector 9,5 - 10 mm
2× straight tube connector 6 - 8 mm
1× exhaust filter size M
5× purple fitting Ø 2,3 mm
5× blue fitting Ø 3,2 mm
5× yellow fitting Ø 3,2 mm
5× green fitting Ø 1,6 mm
Caution: Exhaust filters and air valves have a service life of 6 months.