µ-Dish 35 mm, Low
MSPP-80136
- Ø:35 mm
- Description:µ-Dish, Low
- Height:7 mm
- Cat. No.:MSPP-80136
- Growth area:3.5 cm²
- Working volume:800 µl
- Actual growth surface:410 mm² (using 400 µl)
A 35 mm imaging dish with a polymer coverslip bottom and low walls for high-end microscopy and micromanipulation.
- Low walls for a large access area during micromanipulation assays
- Image on a microscopy dish with a No. 1.5 ibidi Polymer Coverslip Bottom with the highest optical quality and optimal cell adherence
- Lid equipped with locking feature that minimizes evaporation
- Coating Area of 4.1 cm2 (using 400 µl)
The µ-Dish 35mm, Low with 7 mm walls. The low height makes high numerical apertures of Kohler illumination possible and provides large access for micromanipulation. The lid can be closed to hinder evaporation during long term experiments. Ø 35 mm, low wall (800 μl volume), #1.5 polymer coverslip, tissue culture treated.