Order Entry
Puerto Rico
ContactUsLinkComponent
Type I Collagen
Type I Collagen
  MSPP-50202
 :  
Type I Collagen
  MSPP-50202
 :  50202
 :  

 

  • Size:
    4 x 5 ml
  • Storage Conditions:
    –20° C, avoid multiple freeze and thaw cycles
  • Formulation:
    Supplied in 17.5 mM acetic acid (0.1%)
  • pH:
    3.8
  • Cat. No.:
    MSPP-50202
  • Supplier no.:
    50202

 

 

A high-quality rat tail collagen type I solution for cell culture applications (e.g., 3D gels, scaffolds, and coating).

  • Provides in vivo-like extracellular matrix (ECM) structures
  • Non-pepsinized, native collagen for modelling biological ECM in gel matrices
  • Fast polymerization facilitates optimal cell distribution in 3D gels
  • Tested, found negative for DNA, bacteria, fungi, and mycoplasma
  • Sterile for Cell Culture
  • Sourced from Rat Tail Tendon
  • 3D gelling and 2D coating test in cell culture
  • Optically clear viscous liquid

Type I Collagen is a major structural component of the extracellular matrix (ECM). This fibrous protein is often used in three-dimensional (3D) collagen gels that simulate the in vivo cell environment better than the traditional 2D systems. Collagen I can also be used for coating surfaces, as it can form thin layers for culturing cells. ibidi’s Type I Collagen is a state-of-the-art, purified protein extracted from rat tail tendons without pepsinization. It is specially engineered for ibidi’s cell-based assays (e.g., chemotaxis assays in 3D gels, or 2D coating of lab cultureware).