Mounting Medium with DAPI
Catalog # MSPP-50011
Supplier: IBIDI USA MS
CAS Number:
Specifications
- Description:Mounting Medium with DAPI
- Storage Conditions:Dark
- Refractive Index:1.42 - 1.44
- Storage Temperature:2…8° C
- pH:8.5 - 9.0
- Cat. No.:MSPP-50011
Specifications
About this item
A mounting medium w/ DAPI that is optimized for fluorescence microscopy and ibidi µ-Slides and µ-Dishes.
- Dropper bottle for exact dispensing into channels or open wells
- DAPI counterstaining and mounting combined in one single step
- Brilliant fluorescence signals with low background fluorescence
- Staining Specificity: DNA/RNA (Nucleus)
- Liquid, Optically Clear Appearence
The ibidi Mounting Medium with DAPI is a glycerol-based mounting solution optimized for ibidi µ-Slides, µ-Dishes, and µ-Plates with adherent cells or tissue sections that are immunofluorescent stained. It contains an inhibitor that retards photobleaching of fluorochromes. ibidi Mounting Medium with DAPI is ready-to-use.
Certifications: In compliance with DIN ISO 4796-1 standards.
Delivery: Supplied with screw cap and pouring ring.
Packaging: The bottles are packaged in shrinked-wrapped modules.