Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Mounting Medium

Mounting Medium, 15 mL
Catalog # MSPP-50001
Marketsource
Supplier:   IBIDI USA MS
 :  50001
Mounting Medium
Catalog # MSPP-50001
Supplier:  IBIDI USA MS
Marketsource
CAS Number:  
Mounting Medium

Mounting Medium, 15 mL

CAS Number:  
Price
$184.62
Log in for availability
Each
 
Product Details & Documents

A mounting medium that is optimized for fluorescence microscopy and ibidi µ-Slides and µ-Dishes.

  • Ready-to-use in widefield fluorescence, confocal microscopy, and immunofluorescence assays
  • No autofluorescence – bright fluorescence signals
  • Dropper bottle for exact dispensing into channels or open wells

ibidi Mounting Medium is a glycerol-based, non-hardening mounting medium optimized for fluorescence microscopy. The dropper bottle is ideal for exact dispensing into channels or open wells such as in the ibidi µ-Slides and µ-Dishes. 15 ml

 : In compliance with DIN ISO 4796-1 standards.

 : Supplied with screw cap and pouring ring.

 : The bottles are packaged in shrinked-wrapped modules.

Specifications

Specifications

  • Description:
    Mounting Medium
  • Size:
    15 mL
  • Storage Conditions:
    Dark, Room Temperature
  • Cat. No.:
    MSPP-50001
  • pH:
    8.5 - 9.0
  • Refractive Index:
    1.42 - 1.44

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.