Orders LinkContactUsLinkComponent
24-Well Plates, Caplugs
24-Well Plates, Caplugs
Supplier:   Caplugs
24-Well Plates, Caplugs
Supplier:  Caplugs
Certificates
24-Well Plates, Caplugs
24-Well Plates, Caplugs
4 Options Available Below
Product Details & Documents
24-well plates are gamma-irradiated for sterility and available in both sterile and treated versions.
  • Distinct alpha-numeric well designations
  • Well volume: 3.5 ml
  • Available with or without lid
Available in individually packed units.
Documents
Product Family Options
Plates
Product Information
  • Material
    Well volume
    Color
    Sterile
    Availability
    Price
  • 76520-634
    24-well plates with lid
    24-well plates with lid
    Polystyrene
    3.5 ml
    Natural
    Sterile
    Log in for availability
    Log in for availability
    Carton of 100
    Carton of 100 - $286.18
    $286.18
    Carton of 100
       $286.18
    Supplier Number: 222-8044-01F
  • 76520-642
    24-well plates without lid
    24-well plates without lid
    PP
    3.5 ml
    Natural
    Non sterile
    Log in for availability
    Log in for availability
    Carton of 100
    Carton of 100 - $270.82
    $270.82
    Carton of 100
       $270.82
    Supplier Number: 290-8324-03F
  • 76520-636
    24-well plates without lid
    24-well plates without lid
    Polystyrene
    3.5 ml
    Natural
    Non sterile
    Log in for availability
    Log in for availability
    Carton of 100
    Carton of 100 - $176.73
    $176.73
    Carton of 100
       $176.73
    Supplier Number: 290-8124-01F
  • 76520-644
    24-well plates with lid, TC treated
    24-well plates with lid, TC treated
    Polystyrene
    3.5 ml
    Natural
    Sterile
    Log in for availability
    Log in for availability
    Carton of 100
    Carton of 100 - $424.69
    $424.69
    Carton of 100
       $424.69
    Supplier Number: 333-8024-01F
Lids
Product Information
  • Catalog #
    Material
    Sterility
    Color
    Description
    Availability
    Price
  • Polystyrene
    Non sterile
    Natural
    Lid for 24-well plate
    Log in for availability
    Log in for availability
    Carton of 105
    Carton of 105 - $100.83
    $100.83
    Carton of 105
       $100.83
Product Information
  • Catalog #
    Material
    Sterility
    Color
    Description
    Availability
    Price
  • PP
    Non sterile
    Natural
    Lid for 24-well plate
    Log in for availability
    Log in for availability
    Carton of 105
    Carton of 105 - $88.96
    $88.96
    Carton of 105
       $88.96