Orders LinkContactUsLinkComponent
6-Well Plates, Caplugs
6-Well Plates, Caplugs
Supplier:   Caplugs
6-Well Plates, Caplugs
Supplier:  Caplugs
Certificates
6-Well Plates, Caplugs
6-Well Plates, Caplugs
2 Options Available Below
Product Details & Documents
6-well plates are gamma-irradiated for sterility and available in both sterile and treated versions.
  • Distinct alpha-numeric well designations
  • Well volume: 17.7 ml
  • Lid included with plate
Available in individually packed units.
Product Family Options
Product Information
  • Material
    Well volume
    Color
    Sterile
    Availability
    Price
  • 76520-650
    6-well plates with lid
    6-well plates with lid
    Polystyrene
    17.7 ml
    Natural
    Sterile
    Log in for availability
    Log in for availability
    Carton of 100
    Carton of 100 - $327.37
    $327.37
    Carton of 100
       $327.37
    Supplier Number: 222-8048-01F
  • 76520-652
    6-well plates with lid, TC treated
    6-well plates with lid, TC treated
    Polystyrene
    17.7 ml
    Natural
    Sterile
    Log in for availability
    Log in for availability
    Carton of 50
    Carton of 50 - $191.51
    $191.51
    Carton of 50
       $191.51
    Supplier Number: 333-8028-01F