Order Entry
Puerto Rico
ContactUsLinkComponent
VWR® USB Digital Data Logger (DDL) (Dual Probe)
VWR® USB Digital Data Logger (DDL) (Dual Probe)
Catalog # 76520-154
Supplier:  VWR International
undefined
VWR® USB Digital Data Logger (DDL) (Dual Probe)
Catalog # 76520-154
Supplier:  VWR International

Specifications

  • Description:
    USB digital data logger (DDL) (dual probe)
  • Weight:
    1.1 kg (2.5 lbs.)
  • Exterior W×D×H:
    203×127×76 mm (8×5×3")
  • Cat. No.:
    76520-154

Specifications

About this item

This USB data logger is compliant with current CDC vaccine storage monitor guidelines. It includes a buffered probe in product simulated solution, °F/°C switchable readout, a detachable dual probe, audible and visual alarms,and temperature data downloadable via USB drive (not included). This device stores up to 3 years of data.

  • Material: Assorted materials