VWR® USB digital data logger (DDL) for Ultra Low Temperature Freezers (single probe)
76520-146
- Description:USB digital data logger (DDL), for use with Ultra low temperature freezers (single probe)
- Weight:1.1 kg (2.5 lbs.)
- Exterior W×D×H:203×127×76 mm (8×5×3")
- Cat. No.:76520-146
This USB data logger is compliant with CDC guidelines. It comes with a buffered probe in product simulated solution, °F/°C switchable readout, a detachable single probe, audible and visual alarms, and temperature data downloadable via USB drive (not included). Device stores up to 3 years of data.
- Material: Assorted materials