Autoclave Wrappers for Steam, EtO or Gamma Sterilization, Blue BHD, Keystone Cleanroom Products
Supplier: KEYSTONE ADJUSTABLE CAP CO., INC.
Certificates
About this item
When secured with autoclave tape or elasticized bands, Keystone Cleanroom Product’s Blue BHD wrappers provide an effective microbial barrier from external sources of contamination.
- Designed for steam sterilization, but also compatible with EtO or gamma irradiation
- Treated to impart water and alcohol repellency
- Wet formed, lightweight non-woven fabric comprised of natural wood pulp and polyester fibers bonded with a resin binder
- Superior bacterial barrier properties compared to linen
- Sterilization tapes adhere firmly and remove easily
- Ideal choice for for light trays, small basins and towels
- Resists tearing and punctures
- Does not contain any natural latex rubber
These versatile wraps offer fluid repellency and provide an excellent barrier against airborne bacteria. The soft, drapable non-woven blend of cellulose and polyester is virtually lint-free and helps protect wrapped contents from contamination.
Certifications: Certificates of Conformance with each shipment.
Packaging: Processed and packaged in ISO certified cleanrooms.
Frequently Bought Together