Orders LinkContactUsLinkComponent
Membrane Filter Funnels, SARTORIUS
Membrane Filter Funnels, SARTORIUS
Marketsource
Supplier:   Sartorius
Membrane Filter Funnels, SARTORIUS
Supplier:  Sartorius
Marketsource
Certificates
Membrane Filter Funnels, SARTORIUS
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Stainless steel, autoclavable.
To use with filter holders for colony counting.
Ordering information: Supplied with funnel and clamp (lid and seal must be ordered separately).
Product Family Options
Product Information
  • Capacity
    Availability
    Price
  • 102516-510
    Funnel with clamp
    Funnel with clamp
    500 ml
    Log in for availability
    Log in for availability
    Each
    Each - $512.62
    $512.62
    Each
       $512.62
    Supplier Number: 6981002
    Specifications:
    Marketsource
    Marketsource
  • 102516-520
    Funnel with clamp
    Funnel with clamp
    100 ml
    Log in for availability
    Log in for availability
    Each
    Each - $469.46
    $469.46
    Each
       $469.46
    Supplier Number: 6981065
    Specifications:
    Marketsource
    Marketsource