Vacuum Filter Holders, Sartorius
For colony counting and particle collection, specially designed for applications in which the particles or microorganisms retained on the membrane filter surface are of interest. The stainless steel frit filter support ensures a uniform distribution of the residues.
Autoclavable at +134 °C
Outlet diameter: 10 mm
Composition: with cover, clamp and tap in stainless steel