ContactUsLinkComponent
Nitrogen Generator, NG3000 ultra high purity, Peak Scientific
Nitrogen Generator, NG3000 ultra high purity, Peak Scientific
Catalog # 103549-810
Supplier:  Avantor
Nitrogen Generator, NG3000 ultra high purity, Peak Scientific
Catalog # 103549-810
Supplier:  Avantor
Specifications
  • Description:
    Nitrogen generator, NG3000 ultra high purity
  • Cat. No.:
    103549-810
Specifications
About this item
The NG3000 delivers ultra high purity nitrogen at flows of up to 3 L/min for GC, DSC and DMA applications, as well as other laboratory applications requiring up to 3 L/min of ultra high purity nitrogen.
  • Suitable for applications such as GC, DSC, DMA
  • Suitable for applications requiring ultra high purity
  • Available with or without an external compressor
  • Contains self-regenerating Carbon Molecular Sieve column
  • Gas is supplied on demand so generator works to your schedule
  • 12 month comprehensive on-site warranty
The NG3000 delivers ultra high purity nitrogen at flows of up to 3 L/min for GC, DSC and DMA applications, as well as other laboratory applications requiring up to 3 L/min of ultra high purity nitrogen. The NG series can be supported by in-house air or supplied with an integrated compressor.
To reach their ultra high purity nitrogen output these generators use Pressure Swing Adsorption and a Carbon Molecular Sieve. These separation technologies remove oxygen and other impurities in air, delivering hassle-free, ultra high purity nitrogen, on-demand for the laboratory.