ContactUsLinkComponent
ZORBAX Ion Exchange, Agilent Technologies
ZORBAX Ion Exchange, Agilent Technologies
Certificates
undefined
ZORBAX Ion Exchange, Agilent Technologies
Specifications
  • Catalog No:
  • AG860700-704
  • AG880952-203
  • AG880952-714
  • AG883700-704
  • AG883952-714
  • AG820950-904
  • Max. Pressure:
  • 350 bar
  • 350 bar
  • 350 bar
  • 350 bar
  • 350 bar
  • 350 bar
  • pH Range:
  • 2-7
  • 2-7
  • 2-7
  • 2-7
  • 2-7
  • 2-7
  • Temperature Limit:
  • 80 °C
  • 80 °C
  • 80 °C
  • 80 °C
  • 80 °C
  • 80 °C
  • Wetted Materials:
  • Phosphate buffer
  • Phosphate Buffer
  • Phosphate Buffer
  • Phosphate Buffer
  • Phosphate Buffer
  • Phosphate buffer