Orders LinkContactUsLinkComponent
J&W DB-WAX Columns, Agilent Technologies
J&W DB-WAX Columns, Agilent Technologies
Certificates
J&W DB-WAX Columns, Agilent Technologies
40 Options Available Below
Product Details & Documents
A high-polarity polyethylene glycol (PEG) column that is ideal for food, fragrance, and flavor applications.
  • Polyethylene glycol (PEG) stationary phase
  • Close equivalent to USP Phase G16
  • High polarity
  • Temperature limit of 20 °C is lowest of any bonded PEG phase
  • Improves resolution of low boiling point analytes
  • High column-to-column reproducibility
  • Bonded, crosslinked, and solvent rinsable
  • Exact replacement of HP-WAX
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
Agilent J&W DB-WAX is ideal for food, fragrance, and flavor applications. The column's lower temperature limit of 20 °C improves resolution of analytes with low boiling points.
Specifications for Product Family
Specifications
  • Catalog No:
  • AG127-7012
  • AG127-7013
  • AG127-7022
  • AG127-7023
  • AG127-7023E
  • AG128-7022
  • AG128-7032
  • AG128-7052
  • AG122-7012
  • AG122-7012E
  • AG122-7013
  • AG122-7032
  • AG122-7033
  • AG122-7033E
  • AG122-7032E
  • AG122-7031
  • AG122-7032LTM
  • AG122-7062
  • AG122-7061
  • AG122-7062E
  • AG122-7063
  • AG123-7012
  • AG123-7013
  • AG123-7032
  • AG123-7033
  • AG123-7033E
  • AG123-7032E
  • AG123-7062
  • AG123-7063
  • AG123-7063E
  • AG124-7032
  • AG125-7012
  • AG125-7012E
  • AG125-7017
  • AG125-7032
  • AG125-7032E
  • AG125-7031
  • AG125-7037
  • AG125-7062
  • AG125-7062E
  • Polarity:
  • High-polarity stationary phase
  • High-polarity stationary phase
  • High-polarity stationary phase
  • High-polarity stationary phase
  • High-polarity stationary phase
  • High-polarity stationary phase
  • High-polarity stationary phase
  • High-polarity stationary phase
  • High-polarity stationary phase
  • High-polarity stationary phase
  • High-polarity stationary phase
  • High-polarity stationary phase
  • High-polarity stationary phase
  • High-polarity stationary phase
  • High-polarity stationary phase
  • High-polarity stationary phase
  • High-polarity stationary phase
  • High-polarity stationary phase
  • High-polarity stationary phase
  • High-polarity stationary phase
  • High-polarity stationary phase
  • High-polarity stationary phase
  • High-polarity stationary phase
  • High-polarity stationary phase
  • High-polarity stationary phase
  • High-polarity stationary phase
  • High-polarity stationary phase
  • High-polarity stationary phase
  • High-polarity stationary phase
  • High-polarity stationary phase
  • High-polarity stationary phase
  • High-polarity stationary phase
  • High-polarity stationary phase
  • High-polarity stationary phase
  • High-polarity stationary phase
  • High-polarity stationary phase
  • High-polarity stationary phase
  • High-polarity stationary phase
  • High-polarity stationary phase
  • High-polarity stationary phase
  • USP Code:
  • G14, G15, G16, G20, G39, G47
  • G14, G15, G16, G20, G39, G47
  • G14, G15, G16, G20, G39, G47
  • G14, G15, G16, G20, G39, G47
  • G14, G15, G16, G20, G39, G47
  • G14, G15, G16, G20, G39, G47
  • G14, G15, G16, G20, G39, G47
  • G14, G15, G16, G20, G39, G47
  • G14, G15, G16, G20, G39, G47
  • G14, G15, G16, G20, G39, G47
  • G14, G15, G16, G20, G39, G47
  • G14, G15, G16, G20, G39, G47
  • G14, G15, G16, G20, G39, G47
  • G14, G15, G16, G20, G39, G47
  • G14, G15, G16, G20, G39, G47
  • G14, G15, G16, G20, G39, G47
  • G14, G15, G16, G20, G39, G47
  • G14, G15, G16, G20, G39, G47
  • G14, G15, G16, G20, G39, G47
  • G14, G15, G16, G20, G39, G47
  • G14, G15, G16, G20, G39, G47
  • G14, G15, G16, G20, G39, G47
  • G14, G15, G16, G20, G39, G47
  • G14, G15, G16, G20, G39, G47
  • G14, G15, G16, G20, G39, G47
  • G14, G15, G16, G20, G39, G47
  • G14, G15, G16, G20, G39, G47
  • G14, G15, G16, G20, G39, G47
  • G14, G15, G16, G20, G39, G47
  • G14, G15, G16, G20, G39, G47
  • G14, G15, G16, G20, G39, G47
  • G14, G15, G16, G20, G39, G47
  • G14, G15, G16, G20, G39, G47
  • G14, G15, G16, G20, G39, G47
  • G14, G15, G16, G20, G39, G47
  • G14, G15, G16, G20, G39, G47
  • G14, G15, G16, G20, G39, G47
  • G14, G15, G16, G20, G39, G47
  • G14, G15, G16, G20, G39, G47
  • G14, G15, G16, G20, G39, G47
Specifications
Product Family Options
Product Information
  • Column Format
    Phase
    Column hardware
    Film Thickness
    Ø int.
    Length
    Availability
    Price
  • AG127-7012
    GC column, 7" cage
    GC column, 7" cage
    Capillary
    DB-WAX
    Fused silica
    0.10 µm
    0.10 mm
    10 m
    Log in for availability
    Log in for availability
    Each
    Each - $535.00
    $535.00
    Each
       $535.00
    Supplier Number: 127-7012
    Specifications:
    More
    Specifications
    Cat. No.
    AG127-7012
    Polarity
    High-polarity stationary phase
    USP Code
    G14, G15, G16, G20, G39, G47
  • AG127-7013
    GC column, 7" cage
    GC column, 7" cage
    Capillary
    DB-WAX
    Fused silica
    0.20 µm
    0.10 mm
    10 m
    Log in for availability
    Log in for availability
    Each
    Each - $535.00
    $535.00
    Each
       $535.00
    Supplier Number: 127-7013
    Specifications:
    More
    Specifications
    Cat. No.
    AG127-7013
    Polarity
    High-polarity stationary phase
    USP Code
    G14, G15, G16, G20, G39, G47
  • AG127-7022
    GC column, 7" cage
    GC column, 7" cage
    Capillary
    DB-WAX
    Fused silica
    0.1 µm
    0.10 mm
    20 m
    Log in for availability
    Log in for availability
    Each
    Each - $816.00
    $816.00
    Each
       $816.00
    Supplier Number: 127-7022
    Specifications:
    More
    Specifications
    Cat. No.
    AG127-7022
    Polarity
    High-polarity stationary phase
    USP Code
    G14, G15, G16, G20, G39, G47
  • AG127-7023
    GC column, 7" cage
    GC column, 7" cage
    Capillary
    DB-WAX
    Fused silica
    0.20 µm
    0.10 mm
    20 m
    Log in for availability
    Log in for availability
    Each
    Each - $816.00
    $816.00
    Each
       $816.00
    Supplier Number: 127-7023
    Specifications:
    More
    Specifications
    Cat. No.
    AG127-7023
    Polarity
    High-polarity stationary phase
    USP Code
    G14, G15, G16, G20, G39, G47
  • AG127-7023E
    GC column, 5" cage
    GC column, 5" cage
    Capillary
    DB-WAX
    Fused silica
    0.20 µm
    0.10 mm
    20 m
    Log in for availability
    Log in for availability
    Each
    Each - $917.00
    $917.00
    Each
       $917.00
    Supplier Number: 127-7023E
    Specifications:
    More
    Specifications
    Cat. No.
    AG127-7023E
    Polarity
    High-polarity stationary phase
    USP Code
    G14, G15, G16, G20, G39, G47
  • AG128-7022
    GC column, 7" cage
    GC column, 7" cage
    Capillary
    DB-WAX
    Fused silica
    0.20 µm
    0.20 mm
    25 m
    Log in for availability
    Log in for availability
    Each
    Each - $642.00
    $642.00
    Each
       $642.00
    Supplier Number: 128-7022
    Specifications:
    More
    Specifications
    Cat. No.
    AG128-7022
    Polarity
    High-polarity stationary phase
    USP Code
    G14, G15, G16, G20, G39, G47
  • AG128-7032
    GC column, 7" cage
    GC column, 7" cage
    Capillary
    DB-WAX
    Fused silica
    0.20 µm
    0.20 mm
    30 m
    Log in for availability
    Log in for availability
    Each
    Each - $751.00
    $751.00
    Each
       $751.00
    Supplier Number: 128-7032
    Specifications:
    More
    Specifications
    Cat. No.
    AG128-7032
    Polarity
    High-polarity stationary phase
    USP Code
    G14, G15, G16, G20, G39, G47
  • AG128-7052
    GC column, 7" cage
    GC column, 7" cage
    Capillary
    DB-WAX
    Fused silica
    0.20 µm
    0.20 mm
    50 m
    Log in for availability
    Log in for availability
    Each
    Each - $1,090.00
    $1,090.00
    Each
       $1,090.00
    Supplier Number: 128-7052
    Specifications:
    More
    Specifications
    Cat. No.
    AG128-7052
    Polarity
    High-polarity stationary phase
    USP Code
    G14, G15, G16, G20, G39, G47
  • AG122-7012
    GC column, 7" cage
    GC column, 7" cage
    Capillary
    DB-WAX
    Fused silica
    0.25 µm
    0.25 mm
    15 m
    Log in for availability
    Log in for availability
    Each
    Each - $466.00
    $466.00
    Each
       $466.00
    Supplier Number: 122-7012
    Specifications:
    More
    Specifications
    Cat. No.
    AG122-7012
    Polarity
    High-polarity stationary phase
    USP Code
    G14, G15, G16, G20, G39, G47
  • AG122-7012E
    GC column, 5" cage
    GC column, 5" cage
    Capillary
    DB-WAX
    Fused silica
    0.25 µm
    0.25 mm
    15 m
    Log in for availability
    Log in for availability
    Each
    Each - $519.00
    $519.00
    Each
       $519.00
    Supplier Number: 122-7012E
    Specifications:
    More
    Specifications
    Cat. No.
    AG122-7012E
    Polarity
    High-polarity stationary phase
    USP Code
    G14, G15, G16, G20, G39, G47
  • AG122-7013
    GC column, 7" cage
    GC column, 7" cage
    Capillary
    DB-WAX
    Fused silica
    0.50 µm
    0.25 mm
    15 m
    Log in for availability
    Log in for availability
    Each
    Each - $466.00
    $466.00
    Each
       $466.00
    Supplier Number: 122-7013
    Specifications:
    More
    Specifications
    Cat. No.
    AG122-7013
    Polarity
    High-polarity stationary phase
    USP Code
    G14, G15, G16, G20, G39, G47
  • AG122-7032
    GC column, 7" cage
    GC column, 7" cage
    Capillary
    DB-WAX
    Fused silica
    0.25 µm
    0.25 mm
    30 m
    Log in for availability
    Log in for availability
    Each
    Each - $785.00
    $785.00
    Each
       $785.00
    Supplier Number: 122-7032
    Specifications:
    More
    Specifications
    Cat. No.
    AG122-7032
    Polarity
    High-polarity stationary phase
    USP Code
    G14, G15, G16, G20, G39, G47
  • AG122-7033
    GC column, 7" cage
    GC column, 7" cage
    Capillary
    DB-WAX
    Fused silica
    0.50 µm
    0.25 mm
    30 m
    Log in for availability
    Log in for availability
    Each
    Each - $785.00
    $785.00
    Each
       $785.00
    Supplier Number: 122-7033
    Specifications:
    More
    Specifications
    Cat. No.
    AG122-7033
    Polarity
    High-polarity stationary phase
    USP Code
    G14, G15, G16, G20, G39, G47
  • AG122-7033E
    GC column, 5" cage
    GC column, 5" cage
    Capillary
    DB-WAX
    Fused silica
    0.50 µm
    0.25 mm
    30 m
    Log in for availability
    Log in for availability
    Each
    Each - $876.00
    $876.00
    Each
       $876.00
    Supplier Number: 122-7033E
    Specifications:
    More
    Specifications
    Cat. No.
    AG122-7033E
    Polarity
    High-polarity stationary phase
    USP Code
    G14, G15, G16, G20, G39, G47
  • AG122-7032E
    GC column, 5" cage
    GC column, 5" cage
    Capillary
    DB-WAX
    Fused silica
    0.25 µm
    0.25 mm
    30 m
    Log in for availability
    Log in for availability
    Each
    Each - $876.00
    $876.00
    Each
       $876.00
    Supplier Number: 122-7032E
    Specifications:
    More
    Specifications
    Cat. No.
    AG122-7032E
    Polarity
    High-polarity stationary phase
    USP Code
    G14, G15, G16, G20, G39, G47
  • AG122-7031
    GC column, 7" cage
    GC column, 7" cage
    Capillary
    DB-WAX
    Fused silica
    0.15 µm
    0.25 mm
    30 m
    Log in for availability
    Log in for availability
    Each
    Each - $785.00
    $785.00
    Each
       $785.00
    Supplier Number: 122-7031
    Specifications:
    More
    Specifications
    Cat. No.
    AG122-7031
    Polarity
    High-polarity stationary phase
    USP Code
    G14, G15, G16, G20, G39, G47
  • AG122-7032LTM
    GC column, LTM column module
    GC column, LTM column module
    Capillary
    DB-WAX
    Fused silica
    0.25 µm
    0.25 mm
    30 m
    Log in for availability
    Log in for availability
    Each
    Each - $1,910.00
    $1,910.00
    Each
       $1,910.00
    Supplier Number: 122-7032LTM
    Specifications:
    More
    Specifications
    Cat. No.
    AG122-7032LTM
    Polarity
    High-polarity stationary phase
    USP Code
    G14, G15, G16, G20, G39, G47
  • AG122-7062
    GC column, 7" cage
    GC column, 7" cage
    Capillary
    DB-WAX
    Fused silica
    0.25 µm
    0.25 mm
    60 m
    Log in for availability
    Log in for availability
    Each
    Each - $1,380.00
    $1,380.00
    Each
       $1,380.00
    Supplier Number: 122-7062
    Specifications:
    More
    Specifications
    Cat. No.
    AG122-7062
    Polarity
    High-polarity stationary phase
    USP Code
    G14, G15, G16, G20, G39, G47
  • AG122-7061
    GC column, 7" cage
    GC column, 7" cage
    Capillary
    DB-WAX
    Fused silica
    0.15 µm
    0.25 mm
    60 m
    Log in for availability
    Log in for availability
    Each
    Each - $1,380.00
    $1,380.00
    Each
       $1,380.00
    Supplier Number: 122-7061
    Specifications:
    More
    Specifications
    Cat. No.
    AG122-7061
    Polarity
    High-polarity stationary phase
    USP Code
    G14, G15, G16, G20, G39, G47
  • AG122-7062E
    GC column, 5" cage
    GC column, 5" cage
    Capillary
    DB-WAX
    Fused silica
    0.25 µm
    0.25 mm
    60 m
    Log in for availability
    Log in for availability
    Each
    Each - $1,550.00
    $1,550.00
    Each
       $1,550.00
    Supplier Number: 122-7062E
    Specifications:
    More
    Specifications
    Cat. No.
    AG122-7062E
    Polarity
    High-polarity stationary phase
    USP Code
    G14, G15, G16, G20, G39, G47
  • AG122-7063
    GC column, 7" cage
    GC column, 7" cage
    Capillary
    DB-WAX
    Fused silica
    0.50 µm
    0.25 mm
    60 m
    Log in for availability
    Log in for availability
    Each
    Each - $1,380.00
    $1,380.00
    Each
       $1,380.00
    Supplier Number: 122-7063
    Specifications:
    More
    Specifications
    Cat. No.
    AG122-7063
    Polarity
    High-polarity stationary phase
    USP Code
    G14, G15, G16, G20, G39, G47
  • AG123-7012
    GC column, 7" cage
    GC column, 7" cage
    Capillary
    DB-WAX
    Fused silica
    0.25 µm
    0.32 mm
    15 m
    Log in for availability
    Log in for availability
    Each
    Each - $505.00
    $505.00
    Each
       $505.00
    Supplier Number: 123-7012
    Specifications:
    More
    Specifications
    Cat. No.
    AG123-7012
    Polarity
    High-polarity stationary phase
    USP Code
    G14, G15, G16, G20, G39, G47
  • AG123-7013
    GC column, 7" cage
    GC column, 7" cage
    Capillary
    DB-WAX
    Fused silica
    0.50 µm
    0.32 mm
    15 m
    Log in for availability
    Log in for availability
    Each
    Each - $505.00
    $505.00
    Each
       $505.00
    Supplier Number: 123-7013
    Specifications:
    More
    Specifications
    Cat. No.
    AG123-7013
    Polarity
    High-polarity stationary phase
    USP Code
    G14, G15, G16, G20, G39, G47
  • AG123-7032
    GC column, 7" cage
    GC column, 7" cage
    Capillary
    DB-WAX
    Fused silica
    0.25 µm
    0.32 mm
    30 m
    Log in for availability
    Log in for availability
    Each
    Each - $846.00
    $846.00
    Each
       $846.00
    Supplier Number: 123-7032
    Specifications:
    More
    Specifications
    Cat. No.
    AG123-7032
    Polarity
    High-polarity stationary phase
    USP Code
    G14, G15, G16, G20, G39, G47
  • AG123-7033
    GC column, 7" cage
    GC column, 7" cage
    Capillary
    DB-WAX
    Fused silica
    0.50 µm
    0.32 mm
    30 m
    Log in for availability
    Log in for availability
    Each
    Each - $846.00
    $846.00
    Each
       $846.00
    Supplier Number: 123-7033
    Specifications:
    More
    Specifications
    Cat. No.
    AG123-7033
    Polarity
    High-polarity stationary phase
    USP Code
    G14, G15, G16, G20, G39, G47
  • AG123-7033E
    GC column, 5" cage
    GC column, 5" cage
    Capillary
    DB-WAX
    Fused silica
    0.50 µm
    0.32 mm
    30 m
    Log in for availability
    Log in for availability
    Each
    Each - $949.00
    $949.00
    Each
       $949.00
    Supplier Number: 123-7033E
    Specifications:
    More
    Specifications
    Cat. No.
    AG123-7033E
    Polarity
    High-polarity stationary phase
    USP Code
    G14, G15, G16, G20, G39, G47
  • AG123-7032E
    GC column, 5" cage
    GC column, 5" cage
    Capillary
    DB-WAX
    Fused silica
    0.25 µm
    0.32 mm
    30 m
    Log in for availability
    Log in for availability
    Each
    Each - $949.00
    $949.00
    Each
       $949.00
    Supplier Number: 123-7032E
    Specifications:
    More
    Specifications
    Cat. No.
    AG123-7032E
    Polarity
    High-polarity stationary phase
    USP Code
    G14, G15, G16, G20, G39, G47
  • AG123-7062
    GC column, 7" cage
    GC column, 7" cage
    Capillary
    DB-WAX
    Fused silica
    0.25 µm
    0.32 mm
    60 m
    Log in for availability
    Log in for availability
    Each
    Each - $1,500.00
    $1,500.00
    Each
       $1,500.00
    Supplier Number: 123-7062
    Specifications:
    More
    Specifications
    Cat. No.
    AG123-7062
    Polarity
    High-polarity stationary phase
    USP Code
    G14, G15, G16, G20, G39, G47
  • AG123-7063
    GC column, 7" cage
    GC column, 7" cage
    Capillary
    DB-WAX
    Fused silica
    0.50 µm
    0.32 mm
    60 m
    Log in for availability
    Log in for availability
    Each
    Each - $1,500.00
    $1,500.00
    Each
       $1,500.00
    Supplier Number: 123-7063
    Specifications:
    More
    Specifications
    Cat. No.
    AG123-7063
    Polarity
    High-polarity stationary phase
    USP Code
    G14, G15, G16, G20, G39, G47
  • AG123-7063E
    GC column, 5" cage
    GC column, 5" cage
    Capillary
    DB-WAX
    Fused silica
    0.50 µm
    0.32 mm
    60 m
    Log in for availability
    Log in for availability
    Each
    Each - $1,680.00
    $1,680.00
    Each
       $1,680.00
    Supplier Number: 123-7063E
    Specifications:
    More
    Specifications
    Cat. No.
    AG123-7063E
    Polarity
    High-polarity stationary phase
    USP Code
    G14, G15, G16, G20, G39, G47
  • AG124-7032
    GC column, 7" cage
    GC column, 7" cage
    Capillary
    DB-WAX
    Fused silica
    0.85 µm
    0.45 mm
    30 m
    Log in for availability
    Log in for availability
    Each
    Each - $939.00
    $939.00
    Each
       $939.00
    Supplier Number: 124-7032
    Specifications:
    More
    Specifications
    Cat. No.
    AG124-7032
    Polarity
    High-polarity stationary phase
    USP Code
    G14, G15, G16, G20, G39, G47
  • AG125-7012
    GC column, 7" cage
    GC column, 7" cage
    Capillary
    DB-WAX
    Fused silica
    1.00 µm
    0.53 mm
    15 m
    Log in for availability
    Log in for availability
    Each
    Each - $519.00
    $519.00
    Each
       $519.00
    Supplier Number: 125-7012
    Specifications:
    More
    Specifications
    Cat. No.
    AG125-7012
    Polarity
    High-polarity stationary phase
    USP Code
    G14, G15, G16, G20, G39, G47
  • AG125-7012E
    GC column, 5" cage
    GC column, 5" cage
    Capillary
    DB-WAX
    Fused silica
    1.00 µm
    0.53 mm
    15 m
    Log in for availability
    Log in for availability
    Each
    Each - $581.00
    $581.00
    Each
       $581.00
    Supplier Number: 125-7012E
    Specifications:
    More
    Specifications
    Cat. No.
    AG125-7012E
    Polarity
    High-polarity stationary phase
    USP Code
    G14, G15, G16, G20, G39, G47
  • AG125-7017
    GC column, 7" cage
    GC column, 7" cage
    Capillary
    DB-WAX
    Fused silica
    0.50 µm
    0.53 mm
    15 m
    Log in for availability
    Log in for availability
    Each
    Each - $519.00
    $519.00
    Each
       $519.00
    Supplier Number: 125-7017
    Specifications:
    More
    Specifications
    Cat. No.
    AG125-7017
    Polarity
    High-polarity stationary phase
    USP Code
    G14, G15, G16, G20, G39, G47
  • AG125-7032
    GC column, 7" cage
    GC column, 7" cage
    Capillary
    DB-WAX
    Fused silica
    1.00 µm
    0.53 mm
    30 m
    Log in for availability
    Log in for availability
    Each
    Each - $939.00
    $939.00
    Each
       $939.00
    Supplier Number: 125-7032
    Specifications:
    More
    Specifications
    Cat. No.
    AG125-7032
    Polarity
    High-polarity stationary phase
    USP Code
    G14, G15, G16, G20, G39, G47
  • AG125-7032E
    GC column, 5" cage
    GC column, 5" cage
    Capillary
    DB-WAX
    Fused silica
    1.00 µm
    0.53 mm
    30 m
    Log in for availability
    Log in for availability
    Each
    Each - $1,060.00
    $1,060.00
    Each
       $1,060.00
    Supplier Number: 125-7032E
    Specifications:
    More
    Specifications
    Cat. No.
    AG125-7032E
    Polarity
    High-polarity stationary phase
    USP Code
    G14, G15, G16, G20, G39, G47
  • AG125-7031
    GC column, 7" cage
    GC column, 7" cage
    Capillary
    DB-WAX
    Fused silica
    0.25 µm
    0.53 mm
    30 m
    Log in for availability
    Log in for availability
    Each
    Each - $939.00
    $939.00
    Each
       $939.00
    Supplier Number: 125-7031
    Specifications:
    More
    Specifications
    Cat. No.
    AG125-7031
    Polarity
    High-polarity stationary phase
    USP Code
    G14, G15, G16, G20, G39, G47
  • AG125-7037
    GC column, 7" cage
    GC column, 7" cage
    Capillary
    DB-WAX
    Fused silica
    0.50 µm
    0.53 mm
    30 m
    Log in for availability
    Log in for availability
    Each
    Each - $939.00
    $939.00
    Each
       $939.00
    Supplier Number: 125-7037
    Specifications:
    More
    Specifications
    Cat. No.
    AG125-7037
    Polarity
    High-polarity stationary phase
    USP Code
    G14, G15, G16, G20, G39, G47
  • AG125-7062
    GC column, 7" cage
    GC column, 7" cage
    Capillary
    DB-WAX
    Fused silica
    1.00 µm
    0.53 mm
    60 m
    Log in for availability
    Log in for availability
    Each
    Each - $1,510.00
    $1,510.00
    Each
       $1,510.00
    Supplier Number: 125-7062
    Specifications:
    More
    Specifications
    Cat. No.
    AG125-7062
    Polarity
    High-polarity stationary phase
    USP Code
    G14, G15, G16, G20, G39, G47
  • AG125-7062E
    GC column, 5" cage
    GC column, 5" cage
    Capillary
    DB-WAX
    Fused silica
    1.00 µm
    0.53 mm
    60 m
    Log in for availability
    Log in for availability
    Each
    Each - $1,690.00
    $1,690.00
    Each
       $1,690.00
    Supplier Number: 125-7062E
    Specifications:
    More
    Specifications
    Cat. No.
    AG125-7062E
    Polarity
    High-polarity stationary phase
    USP Code
    G14, G15, G16, G20, G39, G47