J&W High Efficiency Columns, Agilent Technologies
A simple and cost-effective way to increase sample throughput with existing GC or GC/MS instruments.
- Increase sample throughput and productivity while maintaining resolution
- Reduce carrier gas usage and cost per analysis
- Apply with existing GC and GC/MS instrument
- Operates with He or H2 carrier gas
- Available in more than 20 stationary phases
- Rapid and easy method translation
- Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
- Performance Summary shipped with each column
Agilent High Efficiency GC columns are ideal for applications that require faster run times, such as high-throughput screening, fast process monitoring, and fast method development. They can reduce sample run time by 50% or more without compromising resolution.
Unlike 0.1 mm id columns, Agilent 0.15 and 0.18 mm id columns are compatible with all standard-pressure capillary GC and GC/MS instruments—without requiring expensive high-pressure modifications.
- Catalog No:
- AG121-1324
- AG121-1544
- AG121-9627
- AGCP7340
- AGCP7461
- AGCP7686
- AG121-1524
- AG121-7022
- AGCP7411
- AG121-9723
- Polarity:
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Low-polarity stationary phase
- Mid-polarity stationary phase
- Low-polarity stationary phase
- High-polarity stationary phase
- Mid-polarity stationary phase
- High-polarity stationary phase
- Mid-polarity stationary phase
- -
- USP Code:
- G43
- G51
- G51
- G14, G15, G16, G20, G39, G47