Orders LinkContactUsLinkComponent
Stainless Steel Probes, Electron Microscopy Sciences
Stainless Steel Probes, Electron Microscopy Sciences
Certificates
Stainless Steel Probes, Electron Microscopy Sciences
Stainless Steel Probes, Electron Microscopy Sciences
8 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Stainless steel probes are perfect for lab applications and electronics inspections.
  • 155 mm overall length
  • Multiple tip styles available
Stainless steel probes are made of rust-free stainless steel and have a serrated handle for secure grip.
Product Family Options
Product Information
  • Length
    Tip style
    Availability
    Price
  • 76499-620
    Probe kit
    Probe kit
    155 mm
    Straigth, angled, single bend, hook
    Log in for availability
    Log in for availability
    Each
    Each - $21.22
    $21.22
    Each
       $21.22
    Supplier Number: 78325-91
  • 76499-624
    Probe kit
    Probe kit
    155 mm
    Straigth, angled, single bend, hook, triple bend, flat
    Log in for availability
    Log in for availability
    Each
    Each - $33.80
    $33.80
    Each
       $33.80
    Supplier Number: 78325-93
  • 76499-628
    Probe with straight needle tip
    Probe with straight needle tip
    155 mm
    Straight
    Log in for availability
    Log in for availability
    Each
    Each - $8.00
    $8.00
    Each
       $8.00
    Supplier Number: 78326-06
  • 76499-630
    Probe with angled needle tip
    Probe with angled needle tip
    155 mm
    Angled
    Log in for availability
    Log in for availability
    Each
    Each - $8.00
    $8.00
    Each
       $8.00
    Supplier Number: 78326-07
  • 76499-632
    Probe with single bend tip
    Probe with single bend tip
    155 mm
    Single bend
    Log in for availability
    Log in for availability
    Each
    Each - $8.00
    $8.00
    Each
       $8.00
    Supplier Number: 78326-08
  • 76499-634
    Probe with hook tip
    Probe with hook tip
    155 mm
    Hook
    Log in for availability
    Log in for availability
    Each
    Each - $8.00
    $8.00
    Each
       $8.00
    Supplier Number: 78326-09
  • 76499-636
    Probe with triple bend tip
    Probe with triple bend tip
    155 mm
    Trible bend
    Log in for availability
    Log in for availability
    Each
    Each - $8.00
    $8.00
    Each
       $8.00
    Supplier Number: 78326-10
  • 76499-638
    Probe with flat tip
    Probe with flat tip
    155 mm
    Flat
    Log in for availability
    Log in for availability
    Each
    Each - $8.00
    $8.00
    Each
       $8.00
    Supplier Number: 78326-11