Virus Recombinant Ebola SUDV Trimeric GP (from Insect Cells)
Supplier: ReVacc Scientific
Certificates
About this item
A recombinant Ebola SUDV GPΔmuc protein (derived from strain Boniface) consists of resides 33-313 and 473-637 (MLD residues 314-472 and transmembrane domain residues 638-676 deleted) and was produced by insect S2 cells, followed by purification.
- Protein function: Bind to its receptor
This product contains C-terminal trimerization motif, enterokinase cleavage, a TwinStrep tag and 6 hexa-histidine tag.
This product has been run on SDS-PAGE with reduced condition and native blue gel. The binding of HIV negative control mAb and one published antibody CA45 (Zhao, et al, Cell 2017, 169) was tested in ELISAs.
Specifications
- Source:
- Species:
- Environmentally Preferable: