ContactUsLinkComponent
Orum International TRIO.BAS™ Satellite with Tri-Clamp Connection, Petri Plate Format
Orum International TRIO.BAS™ Satellite with Tri-Clamp Connection, Petri Plate Format
Catalog # 76514-148
Supplier:  Hardy Diagnostics
undefined
Orum International TRIO.BAS™ Satellite with Tri-Clamp Connection, Petri Plate Format
Catalog # 76514-148
Supplier:  Hardy Diagnostics
Supplier Number:  BAS321K
Specifications
  • Description:
    TRIO.BAS™ satellite with Tri-Clamp connection
  • Cat. No.:
    76514-148
  • Head type:
    Petri plate
Specifications
About this item
A customizable air sampling satellite option for use with RABS ISOLATOR or MULTIFLEX instruments.
  • Ideal for use in cleanroom barrier systems where space is limited
Note: Third image displays the Tri-Clamp satellite shown in a horizontal position with stainless steel AISI 316 piping connection and MULTIFLEX 1 instrument (both sold separately).