Orum International TRIO.BAS™ Satellite with Tri-Clamp Connection, Contact Plate Format
Catalog # 76514-146
Supplier: Hardy Diagnostics
Specifications
- Description:TRIO.BAS™ satellite with Tri-Clamp connection
- Cat. No.:76514-146
- Head type:Contact plate
Specifications
About this item
A customizable air sampling satellite option for use with RABS ISOLATOR or MULTIFLEX instruments.
- Ideal for use in cleanroom barrier systems where space is limited
Note: Third image displays the Tri-Clamp Satellite shown in a horizontal position with stainless steel AISI 316 piping connection and MULTIFLEX 1 instrument (both sold separately).