Order Entry
Puerto Rico
ContactUsLinkComponent
Universal Thread Adapter, Bürkle
Universal Thread Adapter, Bürkle
  76501-390
undefined
Universal Thread Adapter, Bürkle
  76501-390
 :  5601-0120

 

  • Description:
    Universal thread adapter
  • Material:
    Stainless steel
  • Cat. No.:
    76501-390
  • Fits tubing O.D.:
    32 mm (1.26")
  • Hole size:
    48 - 75 mm (1.89 - 2.95")

 

 

Universal thread adapter for nearly all drums and other containers with opening of 48 to 75 mm (1.89 to 2.95") in diameter, regardless of the respective thread.

  • Material stainless steel AISI 304 and ePTFE seal
  • Min. height of the container opening 6 mm (0.24"), max. 55 mm (2.17")

The thread adapter can be mounted quickly and easily with clamping screws and can be used for different container openings.

With G2" connector thread for Bürkle drum screw connections and pumps with immersion tube diameter up to 32 mm (1.26").

 : Universal adapter including ePTFE seal and mounting key.