Dosing Device, METTLER TOLEDO®
Catalog # 76307-628
Supplier: Mettler Toledo
Specifications
- Description:EasyPlus dosing device - Easy dose
- Burette Resolution:EasyPlus 50 ml burette (included)
- Cat. No.:76307-628
- Application:Sample preparation
Specifications
About this item
The EasyPlus sample preparation device is designed to perform accurate, error-free, and fully automated, liquid handling tasks down to 10 µl.
- Easy dispensing applications
- Clear controls
- Simple operation
- iTitrate user interface
The instrument automatically prepares your standard solutions. Simply input the weight of your sample and define a target concentration. Designed for simplicity, the Easy Dose will start your routine task when you tap on the home screen. Perform your analysis using either the touch screen or the new responsive USB wheel controller. Enjoy its precise control and comfortable operation.